Learn More
Selleck Chemical LLC ETHYLENE-D4-DIAMINE 2HCL E8219-5MG
SDP
Catalog No. 5000041513
Encompass_Preferred
Ethane-d4-1 2-diamine dihydrochloride is the deuterated form of 1 2-diaminoethane dihydrochloride 1 2-diaminoethane dihydrochloride is a laboratory reagent with protein cross-linking effect mainly used in molecular modification or cell inhibition experiments in biochemical research
Catalog No. 50-000-41513 Supplier Selleck Chemical LLC Supplier No. E82195MG
Ethane-d4-1 2-diamine dihydrochloride is the deuterated form of 1 2-diaminoethane dihydrochloride 1 2-diaminoethane dihydrochloride is a laboratory reagent with protein cross-linking effect mainly used in molecular modification or cell inhibition experiments in biochemical research