Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Diacrylate, Stabilized 70%, Thermo Scientific™

Catalog No. AC275040050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC275040050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC275040050 Supplier Thermo Scientific Chemicals Supplier No. 275040050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2274-11-5
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.16
InChI Key KUDUQBURMYMBIJ-UHFFFAOYSA-N
Synonym ethylene diacrylate, ethylene acrylate, ethylene glycol diacrylate, acrylic acid, ethylene ester, acrylic acid, ethylene glycol diester, 2-propenoic acid, 1,2-ethanediyl ester, acrylic acid, diester with ethylene glycol, 1,2-ethanediyl diacrylate, ethylene glycol dipropenoate, unii-42q902riye
PubChem CID 75282
IUPAC Name 2-prop-2-enoyloxyethyl prop-2-enoate
SMILES C=CC(=O)OCCOC(=O)C=C

Specifications

Color Colorless to Yellow
Density 1.04
Infrared Spectrum Authentic
Refractive Index 1.4520 to 1.4620 (20°C, 589nm)
Quantity 5 g
Beilstein 02, 1469
Stabilizer 100ppm MEHQ
Formula Weight 170.17
Percent Purity ≥65%
Physical Form Liquid
Chemical Name or Material Ethylene diacrylate, 70%
Show More Show Less
Hazard Category LACHRYMATOR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.