Learn More
Apexbio Technology LLC Ethylene dimethanesulfonate 4672-49-5 25mg
SDP
Catalog No. 5000097158
Encompass_Preferred
Ethylene dimethanesulfonate (CAS 4672-49-5) is a methanesulfonate compound known to exert its biological effects through a mild alkylation process Studies indicate that it selectively induces apoptosis in Leydig cells thereby modulating specific cell populations within testicular tissue Due to these properties ethylene dimethanesulfonate serves as a valuable tool in biomedical research for investigating mechanisms of germ cell injury reproductive toxicology and the development of testicular injury models It is also utilized in studies exploring cell signaling pathways and apoptotic mechanisms related to reproductive biology
Catalog No. 50-000-97158 Supplier Apexbio Technology LLC Supplier No. A470025
Ethylene dimethanesulfonate (CAS 4672-49-5) is a methanesulfonate compound known to exert its biological effects through a mild alkylation process Studies indicate that it selectively induces apoptosis in Leydig cells thereby modulating specific cell populations within testicular tissue Due to these properties ethylene dimethanesulfonate serves as a valuable tool in biomedical research for investigating mechanisms of germ cell injury reproductive toxicology and the development of testicular injury models It is also utilized in studies exploring cell signaling pathways and apoptotic mechanisms related to reproductive biology