Learn More
Medchemexpress LLC Ethylene dimethanesulfonate | 4672-49-5 | 98.0% | 10 MM 1 ML
SDP
Catalog No. 5000339692
Encompass_Preferred
Ethylene dimethane sulfonate is a mild alkylating, non-volatile methanesulfonic diester of ethylene glycol, known for its selective pro-apoptotic effects on LCs.
  • Mild alkylating agent
  • Non-volatile methanesulfonic diester of ethylene glycol
  • Exhibits selective pro-apoptotic effects on LCs
  • Disrupts epididymal function, potentially leading to sperm granuloma formation
Catalog No. 50-003-39692 Supplier Medchemexpress LLC Supplier No. HY12952410MM/1ML
Ethylene dimethane sulfonate is a mild alkylating, non-volatile methanesulfonic diester of ethylene glycol, known for its selective pro-apoptotic effects on LCs.
  • Mild alkylating agent
  • Non-volatile methanesulfonic diester of ethylene glycol
  • Exhibits selective pro-apoptotic effects on LCs
  • Disrupts epididymal function, potentially leading to sperm granuloma formation