Learn More
Medchemexpress LLC Ethylene dimethanesulfonate | 4672-49-5 | 98.0% | 25 MG
SDP
Catalog No. 5000341592
Encompass_Preferred
Ethylene dimethanesulfonate is a mild alkylating, non-volatile methanesulfonic diester of ethylene glycol. It has selective pro-apoptotic effects on LCs.
  • Selective pro-apoptotic effects on LCs
  • Mild alkylating agent
  • Non-volatile methanesulfonic diester of ethylene glycol
  • Disrupts epididymal function in rats
  • Leads to sperm granuloma formation in rats
  • Appears as a white to off-white solid
  • Soluble in DMSO
Catalog No. 50-003-41592 Supplier Medchemexpress LLC Supplier No. 25MGHY129524
Ethylene dimethanesulfonate is a mild alkylating, non-volatile methanesulfonic diester of ethylene glycol. It has selective pro-apoptotic effects on LCs.
  • Selective pro-apoptotic effects on LCs
  • Mild alkylating agent
  • Non-volatile methanesulfonic diester of ethylene glycol
  • Disrupts epididymal function in rats
  • Leads to sperm granuloma formation in rats
  • Appears as a white to off-white solid
  • Soluble in DMSO