Learn More
Medchemexpress LLC Ethylene dimethanesulfonate | 4672-49-5 | 98.0% | 50 MG
SDP
Catalog No. 5000340156
Encompass_Preferred
Ethylene dimethanesulfonate is a mild alkylating, non-volatile methanesulfonic diester of ethylene glycol. It demonstrates selective pro-apoptotic effects on LCs.
  • Mild alkylating agent
  • Non-volatile methanesulfonic diester of ethylene glycol
  • Selective pro-apoptotic effects on LCs
Catalog No. 50-003-40156 Supplier Medchemexpress LLC Supplier No. HY12952450MG
Ethylene dimethanesulfonate is a mild alkylating, non-volatile methanesulfonic diester of ethylene glycol. It demonstrates selective pro-apoptotic effects on LCs.
  • Mild alkylating agent
  • Non-volatile methanesulfonic diester of ethylene glycol
  • Selective pro-apoptotic effects on LCs