Learn More
U.S. Pharmacopeia Ethylene Glycol, 107-21-1, MFCD00002885, 0.5mL
Catalog No. NC9786505
Encompass
Molecular formula C2H6O2, Molecular weight 62.07, Melting Point 8.6 °F (-13 °C), Boiling Point 381.2 - 392 °F (194 - 200 °C), Flash Point 231.8 °F (111.0 °C) Closed Cup, Synonyms: Glycol alcohol, 1,2-Ethanediol
Catalog No. NC9786505 Supplier U.S. Pharmacopeia Supplier No. 1265515
Molecular formula C2H6O2, Molecular weight 62.07, Melting Point 8.6 °F (-13 °C), Boiling Point 381.2 - 392 °F (194 - 200 °C), Flash Point 231.8 °F (111.0 °C) Closed Cup, Synonyms: Glycol alcohol, 1,2-Ethanediol