Learn More
Medchemexpress LLC Ethylene glycol | 107-21-1 | 99.9% | 62.07 | 500 G
SDP
Catalog No. 5000365837
Encompass_Preferred
Ethylene glycol is an organic compound utilized as an antifreeze agent and coolant. It exhibits antifreeze activity by forming hydrogen bonds with water molecules, thereby lowering the freezing point. It also serves as a raw material for synthesizing polymers, including polyester fibers and polyurethanes.
  • Reduces freezing point by forming hydrogen bonds with water molecules.
  • Serves as a raw material in the synthesis of polymers.
  • Acts as a solvent and reducing agent in nanomaterial synthesis experiments.
  • Promotes the formation of monodisperse metal nanocrystals.
  • Soluble in DMSO at concentrations of 100 mg/mL or greater.
  • Suitable for research use only.
Catalog No. 50-003-65837 Supplier Medchemexpress LLC Supplier No. 500GHYY0338
Ethylene glycol is an organic compound utilized as an antifreeze agent and coolant. It exhibits antifreeze activity by forming hydrogen bonds with water molecules, thereby lowering the freezing point. It also serves as a raw material for synthesizing polymers, including polyester fibers and polyurethanes.
  • Reduces freezing point by forming hydrogen bonds with water molecules.
  • Serves as a raw material in the synthesis of polymers.
  • Acts as a solvent and reducing agent in nanomaterial synthesis experiments.
  • Promotes the formation of monodisperse metal nanocrystals.
  • Soluble in DMSO at concentrations of 100 mg/mL or greater.
  • Suitable for research use only.