Learn More
Medchemexpress LLC Ethylene glycol | 107-21-1 | MFCD00002885 | 99.9% | 62.07 | 100 G
SDP
Catalog No. 5000364913
Encompass_Preferred
Ethylene glycol is an organic compound often used as an antifreeze agent and coolant. It lowers the freezing point by forming hydrogen bonds with water molecules, thereby exerting antifreeze activity. It is also used as a raw material in the synthesis of polymers such as polyester fibers and polyurethanes.
  • Used as an antifreeze agent and coolant
  • Lowers freezing point by forming hydrogen bonds with water molecules
  • Raw material in the synthesis of polymers (e.g., polyester fibers, polyurethanes)
  • Acts as a solvent and reducing agent in nanomaterial synthesis
  • Promotes formation of monodisperse metal nanocrystals
  • Exhibits cytotoxicity against human HL60 cells (CC50 of 221 μM for 1,2-ethanediol)
  • Exhibits cytotoxicity against human HSC2 cells (CC50 of > 400 μM for 1,2-ethanediol)
  • Exhibits cytotoxicity against human HSC4 cells (CC50 of 385 μM for 1,2-ethanediol)
Catalog No. 50-003-64913 Supplier Medchemexpress LLC Supplier No. 100GHYY0338
Ethylene glycol is an organic compound often used as an antifreeze agent and coolant. It lowers the freezing point by forming hydrogen bonds with water molecules, thereby exerting antifreeze activity. It is also used as a raw material in the synthesis of polymers such as polyester fibers and polyurethanes.
  • Used as an antifreeze agent and coolant
  • Lowers freezing point by forming hydrogen bonds with water molecules
  • Raw material in the synthesis of polymers (e.g., polyester fibers, polyurethanes)
  • Acts as a solvent and reducing agent in nanomaterial synthesis
  • Promotes formation of monodisperse metal nanocrystals
  • Exhibits cytotoxicity against human HL60 cells (CC50 of 221 μM for 1,2-ethanediol)
  • Exhibits cytotoxicity against human HSC2 cells (CC50 of > 400 μM for 1,2-ethanediol)
  • Exhibits cytotoxicity against human HSC4 cells (CC50 of 385 μM for 1,2-ethanediol)