Learn More
Bioworld Ethylene Glycol, 4 L
Catalog No. 50255965
Encompass_Preferred
It is a toxic organic compound used as an automotive antifreeze and a precursor to polymers. It is used as plasticizers in adhesives, lacquers and enamels. It also used as coolant and heat transfer agent. Technical Information:High Purity GradeAcidity: <=0.001%Iron: <=0.0005%Moisture (KF): <=0.5%InChI : 1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2InChIKey : LYCAIKOWRPUZTN-UHFFFAOYSA-NIUPAC_Name: Ethane-1,2-DiolNFPA 704: H-1,F-1,R-0,C-NARTECS Number: KW2975000Synonyms: 1,2-Ethanediol;Glycol;Hypodicarbonous Acid;Monoethylene Glycol;MEG
Catalog No. 50-255-965 Supplier Bioworld Supplier No. 405700042
It is a toxic organic compound used as an automotive antifreeze and a precursor to polymers. It is used as plasticizers in adhesives, lacquers and enamels. It also used as coolant and heat transfer agent. Technical Information:High Purity GradeAcidity: <=0.001%Iron: <=0.0005%Moisture (KF): <=0.5%InChI : 1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2InChIKey : LYCAIKOWRPUZTN-UHFFFAOYSA-NIUPAC_Name: Ethane-1,2-DiolNFPA 704: H-1,F-1,R-0,C-NARTECS Number: KW2975000Synonyms: 1,2-Ethanediol;Glycol;Hypodicarbonous Acid;Monoethylene Glycol;MEG