Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol, 50% v/v, Reagents

Catalog No. 01335365
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-365 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-365 Supplier Reagents Holdings Llc Supplier No. CE24760020F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethylene Glycol, 50% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

pH 10.5 to 11
CAS 7732-18-5, 107-21-1
Color Colorless
Quantity 20 L
Packaging Cubitainer
Physical Form Liquid
Specific Gravity 1.0764
Molecular Formula H2O
Synonym 1,2-Ethanediol, Monoethylene Glycol, Antifreeze
Boiling Point 107°C
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 18.02
Percent Purity 50%
Grade Reagent
Chemical Name or Material Ethylene Glycol, 50% v/v
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.