Learn More
Crescent Chemical Co Inc ETHYLENE GLYCOL BIS(2-AMINOETH
SDP
Catalog No. 50247174
Encompass_Preferred
Ethylene glycol bis(2-aminoethylether)-N,N,N',N'-tetra acetic acid analytical grade High selectivity for Ca2+ over Mg2+. Size - 5G Storage Conditions - +15 °C TO +30 °C Catalog Number - 11290.01 CAS 67-42-5
Catalog No. 50-247-174 Supplier Crescent Chemical Co Inc Supplier No. SE11290.01
Ethylene glycol bis(2-aminoethylether)-N,N,N',N'-tetra acetic acid analytical grade High selectivity for Ca2+ over Mg2+. Size - 5G Storage Conditions - +15 °C TO +30 °C Catalog Number - 11290.01 CAS 67-42-5