Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol-bis-(β-aminoethylether)-N,N,N',N'-tetraacetic Acid, MP Biomedicals™

Catalog No. MP521329380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP521329380 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP521329380 Supplier MP Biomedicals, Inc Supplier No. 0521329380

Ethylene Glycol-bis-(β-aminoethylether)-N,N,N',N'-tetraacetic Acid, White Powder

  • It is useful for the determination of calcium in the presence of magnesium. It can be used to study molecular biology, biochemicals, reagents and chelators.
  • It has been used in a study that illustrated the response of the β-cyclodextrin-hemin complex in the induction of cucumber adventitious rooting through heme oxygenase-1-dependent mechanism and calcium signaling.
  • It is a chelating agent. It is also a buffer component in dideoxy sequencing of RNA using reverse transcriptase.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-42-5
Molecular Formula C14H24N2O10
Molecular Weight (g/mol) 380.35
MDL Number MFCD00004291
InChI Key DEFVIWRASFVYLL-UHFFFAOYSA-N
Synonym Ethyleneglycol-bis(2-aminoethylether)-N,N,N,N-tetraacetic acid,, Egtazic acid, Ethylene-bis(oxyethylenenitrilo)tetraacetic acid, Glycol ether diamine tetraacetic acid
PubChem CID 6207
ChEBI CHEBI:30740

Specifications

Melting Point 241°C (decomposition)(literature)
Color White
Quantity 100 g
Beilstein 1717370
Solubility Information Soluble in 2% alkalline solution (clear, colorless solution).
Physical Form Powder
RTECSNumber AH3760000
storageNote1 Store at Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.