Learn More
Medchemexpress LLC Ethylene glycol bis(succinic acid) | 35415-14-6 | MFCD00045949 | 95.0% | 262.21 g/mol | C10H14O8 | 1 G
SDP
Catalog No. 5000280745
Encompass_Preferred
Ethylene glycol bis(succinic acid) is a biochemical reagent used as a building block and organic compound in life-science research. It is supplied with defined purity for laboratory use and is suitable for synthetic chemistry and biochemical assay applications. The compound has molecular formula C10H14O8 and a molecular weight of 262.21 g/mol.
  • High purity: 95.0%.
  • Available in multiple package sizes for flexible ordering.
  • Suitable for synthesis and biochemical assays.
  • Stable at room temperature; in solvent recommended storage is -20°C (1 year) or -80°C (2 years).
  • Molecular formula C10H14O8 and molecular weight 262.21 g/mol.
Catalog No. 50-002-80745 Supplier Medchemexpress LLC Supplier No. HYW1909541G
Ethylene glycol bis(succinic acid) is a biochemical reagent used as a building block and organic compound in life-science research. It is supplied with defined purity for laboratory use and is suitable for synthetic chemistry and biochemical assay applications. The compound has molecular formula C10H14O8 and a molecular weight of 262.21 g/mol.
  • High purity: 95.0%.
  • Available in multiple package sizes for flexible ordering.
  • Suitable for synthesis and biochemical assays.
  • Stable at room temperature; in solvent recommended storage is -20°C (1 year) or -80°C (2 years).
  • Molecular formula C10H14O8 and molecular weight 262.21 g/mol.