Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol bis(Succinic Acid N-Hydroxysuccinimide Ester), MP Biomedicals™

Catalog No. MP215798291 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215798291 1 g
MP215798283 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP215798291 Supplier MP Biomedicals, Inc Supplier No. 0215798291
Add to Cart
Add to Cart

Ethylene Glycol bis(Succinic Acid N-Hydroxysuccinimide Ester), White Powder

  • It can be used to produce succinic acid 2-[3-(doxorubicin-3'-N-yl)-3-oxopropionyloxy]-ethyl ester N-hydroxysuccinimidyl ester with adriamycin at temperature of 20°C.
  • It is a homobifunctional, cleavable cross-linking reagent. It is typically, coupled to molecules containing primary amines by amide bonds buffered at pH 7.5 (6.5-8.5).
  • The linkage provides a 12-atom spacer.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70539-42-3
Molecular Formula C18H20N2O12
Molecular Weight (g/mol) 456.36
MDL Number MFCD00049057
InChI Key QLHLYJHNOCILIT-UHFFFAOYSA-N
Synonym Ethyleneglycol-bis[succinimidyl succinate], Di(N-succinimidyl) ethyleneglycol disuccinate, EGS, Ethyleneglycol disuccinate di(N-succinimidyl) ester
IUPAC Name 2,5-dioxopyrrolidin-1-yl 1-[2-({4-[(2,5-dioxopyrrolidin-1-yl)oxy]-4-oxobutanoyl}oxy)ethyl] butanedioate
SMILES O=C(CCC(=O)ON1C(=O)CCC1=O)OCCOC(=O)CCC(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 115°C to 133°C
Color Powder
Quantity 1 g
Beilstein 9167119
Solubility Information Soluble in acetic acid: water (1:1): ≤50mg/mL, DMF.
storageNote1 Store at +4°C, Store under nitrogen.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.