Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethylene glycol d6 98 atom5G
Catalog No. 502907563
Encompass
Ethylene glycol-d6 is a deuterated NMR solvent useful in NMR-based research and analyses. Its infrared spectrum has been recorded.
Catalog No. 50-290-7563 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 5305495G
Ethylene glycol-d6 is a deuterated NMR solvent useful in NMR-based research and analyses. Its infrared spectrum has been recorded.