Learn More
Medchemexpress LLC Ethylene glycol diacrylate | 2274-11-5 | MFCD00008629 | 99.4% | 170.16 g/mol | C8H10O4 | 100 MG
SDP
Catalog No. 5000281322
Encompass_Preferred
Ethylene glycol diacrylate is a bifunctional diacrylate monomer used as a cross-linking reagent to produce poly(ethylene glycol) diacrylate and to form crosslinked polymer networks for hydrogels, drug delivery, and materials research. Supplied as a high-purity research-grade reagent with analytical documentation available.
Catalog No. 50-002-81322 Supplier Medchemexpress LLC Supplier No. HYW440548100MG
Ethylene glycol diacrylate is a bifunctional diacrylate monomer used as a cross-linking reagent to produce poly(ethylene glycol) diacrylate and to form crosslinked polymer networks for hydrogels, drug delivery, and materials research. Supplied as a high-purity research-grade reagent with analytical documentation available.