Learn More
Medchemexpress LLC Ethylene glycol dimethacrylate | 97-90-5 | MFCD00008590 | 99.2% | 198.22 g/mol | C10H14O4 | 1 ML
SDP
Catalog No. 5000303148
Encompass_Preferred
Ethylene glycol dimethacrylate is a di-functional methacrylate monomer supplied as a ready-to-use 10 mM solution in DMSO for formulation screening and cross-linking applications. The product is high purity and includes documentation for laboratory use.
  • Ready-to-use 10 mM solution in DMSO, 1 mL volume.
  • High purity suitable for research and materials applications.
  • Effective di-functional monomer for cross-linking and polymerization.
  • Small-volume format for screening and formulation workflows.
  • SDS and COA available for safety and quality verification.
Catalog No. 50-003-03148 Supplier Medchemexpress LLC Supplier No. HYW01494010MM/1ML
Ethylene glycol dimethacrylate is a di-functional methacrylate monomer supplied as a ready-to-use 10 mM solution in DMSO for formulation screening and cross-linking applications. The product is high purity and includes documentation for laboratory use.
  • Ready-to-use 10 mM solution in DMSO, 1 mL volume.
  • High purity suitable for research and materials applications.
  • Effective di-functional monomer for cross-linking and polymerization.
  • Small-volume format for screening and formulation workflows.
  • SDS and COA available for safety and quality verification.