Learn More
Medchemexpress LLC Ethylene glycol dimethacrylate | 97-90-5 | MFCD00008590 | 99.2% | 198.22 g·mol⁻¹ | C10H14O4 | 100 G
SDP
Catalog No. 5000303147
Encompass_Preferred
Ethylene glycol dimethacrylate (EGDMA) is a di-functional methacrylate monomer used as a cross-linking agent in polymer and resin formulations. It is supplied as a colorless to light-yellow liquid with a molecular weight of 198.22 g·mol⁻¹ and high purity, suitable for research and formulation applications including dental materials and photopolymer systems.
  • Di-functional methacrylate monomer for cross-linking
  • Liquid, colorless to light-yellow appearance
  • High purity (≈99%) for laboratory and formulation use
  • Supports polymerization and resin curing applications
  • Compatible with photopolymer and dental resin systems
Catalog No. 50-003-03147 Supplier Medchemexpress LLC Supplier No. HYW014940100G
Ethylene glycol dimethacrylate (EGDMA) is a di-functional methacrylate monomer used as a cross-linking agent in polymer and resin formulations. It is supplied as a colorless to light-yellow liquid with a molecular weight of 198.22 g·mol⁻¹ and high purity, suitable for research and formulation applications including dental materials and photopolymer systems.
  • Di-functional methacrylate monomer for cross-linking
  • Liquid, colorless to light-yellow appearance
  • High purity (≈99%) for laboratory and formulation use
  • Supports polymerization and resin curing applications
  • Compatible with photopolymer and dental resin systems