Learn More
Medchemexpress LLC Ethylene glycol dimethacrylate | 97-90-5 | MFCD00008590 | 99.5% | 198.22 g/mol | C10H14O4 | 50 G
SDP
Catalog No. 5000302419
Encompass_Preferred
Ethylene glycol dimethacrylate (EGDMA) is a methacrylate diester monomer used as a cross-linking agent in polymer and resin formulations. It polymerizes with (meth)acrylate monomers to form cross-linked networks for coatings, adhesives, dental materials, and composite research applications. The material is typically a colorless to light yellow liquid.
  • Acts as a cross-linking monomer to increase polymer network strength.
  • Compatible with free-radical (meth)acrylate polymerization systems.
  • Improves mechanical and thermal properties of cured resins.
  • Available as a high-purity liquid suited for research and materials synthesis.
  • Commonly used in coatings, adhesives, dental materials, and composite formulations.
Catalog No. 50-003-02419 Supplier Medchemexpress LLC Supplier No. HYW01494050G
Ethylene glycol dimethacrylate (EGDMA) is a methacrylate diester monomer used as a cross-linking agent in polymer and resin formulations. It polymerizes with (meth)acrylate monomers to form cross-linked networks for coatings, adhesives, dental materials, and composite research applications. The material is typically a colorless to light yellow liquid.
  • Acts as a cross-linking monomer to increase polymer network strength.
  • Compatible with free-radical (meth)acrylate polymerization systems.
  • Improves mechanical and thermal properties of cured resins.
  • Available as a high-purity liquid suited for research and materials synthesis.
  • Commonly used in coatings, adhesives, dental materials, and composite formulations.