Learn More
Medchemexpress LLC Ethylene glycol dimethacrylate | 97-90-5 | MFCD00008590 | 99.5% | 198.22 g/mol | C10H14O4 | 500 G
SDP
Catalog No. 5000302494
Encompass_Preferred
Ethylene glycol dimethacrylate is a diester monomer used as a cross-linking agent in polymerization and materials research. It contains two methacrylate functional groups that form covalent links between polymer chains, enabling network formation and modification of mechanical and chemical properties.
  • Diester monomer with two methacrylate functional groups.
  • Enables crosslinking in free-radical and other polymerization processes.
  • Improves mechanical strength and solvent resistance of polymer networks.
  • Commonly used in dental materials, coatings, and formulation research.
  • Liquid, colorless to light yellow, suitable for laboratory handling.
  • Molecular formula C10H14O4; molecular weight 198.22 g/mol.
Catalog No. 50-003-02494 Supplier Medchemexpress LLC Supplier No. HYW014940500G
Ethylene glycol dimethacrylate is a diester monomer used as a cross-linking agent in polymerization and materials research. It contains two methacrylate functional groups that form covalent links between polymer chains, enabling network formation and modification of mechanical and chemical properties.
  • Diester monomer with two methacrylate functional groups.
  • Enables crosslinking in free-radical and other polymerization processes.
  • Improves mechanical strength and solvent resistance of polymer networks.
  • Commonly used in dental materials, coatings, and formulation research.
  • Liquid, colorless to light yellow, suitable for laboratory handling.
  • Molecular formula C10H14O4; molecular weight 198.22 g/mol.