Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Ethylene Glycol Dimethacrylate 225 ML
SDP

Catalog No. 50980464
Encompass_Preferred

(Ethylene dimethacrylate)
CAS #97-90-5 C₁₀H₁₄O₄ F.W. 198.22
Boiling point: 66-68°C

Inhibited with 100 ppm HQ. (Hydroquinone)

Catalog No. 50-980-464 Supplier Electron Microscopy Sciences Supplier No. 15300
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Ethylene dimethacrylate)
CAS #97-90-5 C₁₀H₁₄O₄ F.W. 198.22
Boiling point: 66-68°C

Inhibited with 100 ppm HQ. (Hydroquinone)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.