Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene glycol dimethyl ether, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, stabilized

Catalog No. ac442981000
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC442981000 100 mL
AC442980010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC442981000 Supplier Thermo Scientific Chemicals Supplier No. 442981000
Only null left
Add to Cart
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethylene glycol dimethyl ether
Melting Point -69.0°C
CAS 110-71-4
Density 0.8670g/mL
Quantity 100 mL
Flash Point −6°C
Refractive Index 1.3785 to 1.3805
Specific Gravity 0.867
Linear Formula CH3OCH2CH2OCH3
Viscosity 1.1 mPa.s (20°C)
Water 0.005% max.
Synonym monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether
Boiling Point 84.0°C to 86.0°C
Solubility Information Solubility in water: miscible. Other solubilities: solulbe in hydrocarbon solvents, soluble in ethanol, ethyl ether, acetone, carbon, tetrachloride, benzene, dichloroethane
Formula Weight 90.12
Percent Purity 99.5%
Grade Extra Dry over Molecular Sieve
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.