Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, 67-42-5, MFCD00004291, 25 g
Catalog No. NC0881568
Encompass
Linear Formula: (-CH2OCH2CH2N(CH2CO2H)2)2, Molecular Weight: 380.35, for molecular biology, ≥97.0%, Synonym: EGTA, Egtazic acid, Ethylene-bis(oxyethylenenitrilo)tetraacetic acid, Glycol ether diamine tetraacetic acid. A chelating agent useful for the determination of calcium in the presence of magnesium.
Catalog No. NC0881568 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. E388925G
Linear Formula: (-CH2OCH2CH2N(CH2CO2H)2)2, Molecular Weight: 380.35, for molecular biology, ≥97.0%, Synonym: EGTA, Egtazic acid, Ethylene-bis(oxyethylenenitrilo)tetraacetic acid, Glycol ether diamine tetraacetic acid. A chelating agent useful for the determination of calcium in the presence of magnesium.