Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol, For Analysis, Emsure™, MilliporeSigma™

Catalog No. M1096214000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1096214000 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1096214000 Supplier MilliporeSigma Supplier No. 1.09621.4000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

pH 6 to 7.5 (100g/L, H2O, 20°C)
Melting Point 14°C to 10°C
Quantity 4 L
Flash Point 111°C
Packaging Glass Bottle
Vapor Pressure 0.053 hPa (20°C)
Boiling Point 197.6°C (1013 hPa)
Solubility Information 1000g/L
Formula Weight 62.07
Percent Purity >99.5%
Grade Reagent Ph. Eur./Reagent USP
Show More Show Less
EINECSNumber 203-473-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.