Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethylene glycol methyl ether methacrylate

Catalog No. 41533250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-533-250ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-533-250ML Supplier Sigma Aldrich Supplier No. 41533250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Polymers prepared from this monomer may have uses in polyelectrolytes and may be biocompatible.Used in the synthesis of copolymers of styrene and 2-methoxyethyl methacrylate.

Specifications

CAS 6976-93-8
Density 0.993 g/mL (at 25°C)
Boiling Point 65°C (2 mmHg)
Molecular Formula C7H12O3
Refractive Index n20/D 1.431 (literature)
Linear Formula H2C=C(CH3)CO2CH2CH2OCH3
Quantity 50 mL
MDL Number MFCD00048122
Synonym 2-Methoxyethyl methacrylate
Molecular Weight (g/mol) 144.17
Percent Purity 99%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.