Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Ethylene Glycol Monoethyl Ether, Purified 1 LT
SDP

Catalog No. 50980465
Encompass_Preferred

(Cellosolve; 2-Ethoxyethanol; Ethyl Glycol)
CAS #110-80-5 HOCH₂CH₂OCH₂CH₃ F.W. 90.12
Assay: >99.5%
Boiling point: 133-135°C

Suitable for histological use.

Catalog No. 50-980-465 Supplier Electron Microscopy Sciences Supplier No. 1531001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Cellosolve; 2-Ethoxyethanol; Ethyl Glycol)
CAS #110-80-5 HOCH₂CH₂OCH₂CH₃ F.W. 90.12
Assay: >99.5%
Boiling point: 133-135°C

Suitable for histological use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.