Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Ethylene Glycol Monoethyl Ether, Purified 1 LT
SDP

Supplier:  Electron Microscopy Sciences 1531006

Encompass_Preferred

(Cellosolve; 2-Ethoxyethanol; Ethyl Glycol)
CAS #110-80-5 HOCH₂CH₂OCH₂CH₃ F.W. 90.12
Assay: >99.5%
Boiling point: 133-135°C

Suitable for histological use.

Catalog No. 50-980-466


May include imposed supplier surcharges.
Add to Cart
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.