Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol Mono(4-bromophenyl) Ether 98.0+%, TCI America™
SDP

Catalog No. E050425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E050425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E050425G Supplier TCI America Supplier No. E050425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34743-88-9
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.06
MDL Number MFCD00002860
InChI Key QYIOGYCRGNHDNK-UHFFFAOYSA-N
Synonym 2-(4-Bromophenoxy)ethanol
PubChem CID 99168
IUPAC Name 2-(4-bromophenoxy)ethan-1-ol
SMILES OCCOC1=CC=C(Br)C=C1

Specifications

Melting Point 55°C
Color White
Boiling Point 184°C
Quantity 25 g
Formula Weight 217.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethylene Glycol Mono(4-bromophenyl) Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.