Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethylene glycol monohexadecyl ether | 2136-71-2 | MFCD00043350 | ≥97.0% | 286.49 g/mol | C18H38O2 | 25 MG
SDP

Catalog No. 5000281243
Encompass_Preferred

Ethylene glycol monohexadecyl ether is a nonionic surfactant and biochemical assay reagent used to solubilize hydrophobic compounds and improve wetting in laboratory assays. It is provided as a high-purity chemical (CAS 2136-71-2) suitable for research applications, formulation studies, and cell-based assays; handle according to the product safety data sheet.

  • Nonionic surfactant and solubilizing agent.
  • High purity (≥97.0%).
  • Molecular weight 286.49 g/mol.
  • Molecular formula C18H38O2.
  • Available in small research pack sizes (e.g., 25 mg).
  • For research use only; not for human or clinical use.

Catalog No. 50-002-81243 Supplier Medchemexpress LLC Supplier No. HYW25046625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethylene glycol monohexadecyl ether is a nonionic surfactant and biochemical assay reagent used to solubilize hydrophobic compounds and improve wetting in laboratory assays. It is provided as a high-purity chemical (CAS 2136-71-2) suitable for research applications, formulation studies, and cell-based assays; handle according to the product safety data sheet.

  • Nonionic surfactant and solubilizing agent.
  • High purity (≥97.0%).
  • Molecular weight 286.49 g/mol.
  • Molecular formula C18H38O2.
  • Available in small research pack sizes (e.g., 25 mg).
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.