Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethylene glycol phenyl ether acrylate

Catalog No. 4083361L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-833-61L 1 L
40-833-6250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-833-61L Supplier Sigma Aldrich Supplier No. 4083361L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethylene glycol phenyl ether acrylate (PA) is a monofunctional acrylate.Ethylene glycol phenyl ether acrylate (PA) monomer may be used to fabricate polymerization induced grating pattern on photopolymer films.

Specifications

CAS 48145-04-6
Density 1.104 g/mL (at 25°C)
Boiling Point 84°C (0.2 mmHg)
Molecular Formula C11H12O3
Refractive Index n20/D 1.518 (literature)
Linear Formula H2C=CHCO2CH2CH2OC6H5
MDL Number MFCD00053696
Quantity 1 L
Synonym 2-Phenoxyethyl acrylate
Molecular Weight (g/mol) 192.21
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.