Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol, puriss. p.a., Reag. Ph. Eur., ≥99.5%, Solstice

Catalog No. 60044523 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
5 L
6 x 1 L
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-523 250 mL Poly Bottle
60-044-522 1 L Poly Bottle
60-044-525 6 x 1 L Poly Bottle
60-044-524 5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-044-523 Supplier Solstice Supplier No. 03750250ML
Add to Cart
Add to Cart

Puriss. p.a., Reag. Ph. Eur.,≥99.5%

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

pH Neutral
Melting Point -12°C
Color Colorless
Density 1.113g/mL at 25°C
Quantity 250 mL
Explosion Limit 0.153
Flash Point 111°C (231.8°F)
Ignition Point 752°F
Packaging Poly Bottle
Physical Form Liquid
Refractive Index n20/D 1.431
Linear Formula HOCH2CH2OH
Vapor Pressure 0.08mmHg (20°C)
UN Number UN3082
Beilstein 505945
Boiling Point 197°C
Formula Weight 62.07g/mol
Percent Purity ≥99.5%
Grade puriss. p.a.
Chemical Name or Material Ethylene Glycol, Reag. Ph. Eur.
Show More Show Less
Hazard Statement P301 + P312 + P330
Precautionary Statement H302-H373
DOTInformation 9/PGIII
EINECSNumber 203-473-3
RTECSNumber KW2975000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.