Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol, Reagent Grade, Ricca Chemical
SDP

Catalog No. R294450020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R294450020F 20 L
R2944500500 500 mL
R29445004C 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R294450020F Supplier Ricca Chemical Company Supplier No. R294450020F
Add to Cart
Add to Cart

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.07
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES OCCO

Specifications

CAS Max % 100
Appearance Colorless, Viscous Liquid
Quantity 20 L
Packaging Cubitainer
Boiling Point Approximately 197°C
Grade Reagent
Chemical Name or Material Ethylene Glycol
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.