Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene glycol, Reagent Grade, ≥99%, Honeywell™

Catalog No. 60044642 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
20 L
30 kg
4 x 2.5 L
225 kg
Packaging:
Metal Drum
Poly Bottle
Stainless Steel Can
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-642 20 L Stainless Steel Can
60-044-640 1 L Poly Bottle
60-044-647 5 L Poly Bottle
60-044-645 4 x 2.5 L Poly Bottle
60-044-644 30 kg Stainless Steel Can
60-044-643 225 kg Metal Drum
60-044-641 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 60-044-642 Supplier Honeywell Chemicals Supplier No. 10246620L
Only null left
Add to Cart
Add to Cart

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point -13°C (literature)
CAS Max % 100.0000%
Density 1.113 g/mL at 25 °C(lit.)
Quantity 20 L
Flash Point 111°C
Packaging Stainless Steel Can
Linear Formula HOCH2CH2OH
Beilstein 505945
Boiling Point 197°C
Formula Weight 62.07
Percent Purity >99%
Grade Reagent
Chemical Name or Material Ethylene glycol, Reagent Grade, ≥99%
Show More Show Less
ShelfLife 1080 days from date of manufacture
EINECSNumber 203-473-3
Recommended Storage Room Temp
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.