Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol, Technical, Solstice

Catalog No. 60044527 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
5 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-527 5 L Poly Bottle
60-044-526 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-044-527 Supplier Solstice Supplier No. 037705L

missing translation for 'validMainframeMsg'

Technical, suitable as anti-freezing agent (with corrosion-inhibitor), blue

  • Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
  • Variation in color and physical form is possible
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point -20°C
Color Blue
Density 1.113g/mL at 25°C
Quantity 5 L
Explosion Limit 0.153
Flash Point 111°C (231.8°F)
Ignition Point 752°F
Packaging Poly Bottle
Physical Form Liquid
Refractive Index n20/D 1.431(lit.)
Vapor Pressure 0.08mmHg (20°C)
Linear Formula HOCH2CH2OH
UN Number UN3082
Beilstein 505945
Boiling Point 195°C to 198°C
Formula Weight 62.07g/mol
Grade Technical
Chemical Name or Material Ethylene Glycol
Show More Show Less
Hazard Statement P260-P301 + P312 + P330
Precautionary Statement H302-H373
DOTInformation 9/PGIII
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.