Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALADDIN SCIENTIFIC CORPORATION Ethylene imine polymer, 25g

Catalog No. 5000082568
Encompass_Preferred

Ethylene imine polymer 99% M W 10000Cas-9002-98-6

Catalog No. 50-000-82568 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. E10707925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethylene imine polymer 99% M W 10000Cas-9002-98-6

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.