Learn More
3M™ Ethylene Oxide Monitor 3551+
Catalog No. 03052856 Shop All 3M Products
Change view
Click to view available options
Detectable Analytes:
Ethylene Oxide
Catalog No. Detectable Analytes
03-052-856 Ethylene Oxide
1 options
Catalog No. 03-052-856 Supplier 3M™ Supplier No. 7100249356
  • $347.00
    $267.65 / Case of 5
    Save $79.35
    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
3M™ Ethylene Oxide Monitor is a cost-effective and convenient way to perform personal air sampling for ethylene oxide. No batteries, hoses or pumps or needed to use the monitor to sample your air.
  • State of the Art Monitor Design
  • Simple, cost-effective and convenient
  • A valid method for air sampling and analysis of ethylene oxide
  • Easy to use. Just clip the monitor to shirt lapel, collar, or pocket
  • Unobtrusive. Monitors are small and lightweight and will not interfere with employee activities.
  • Documentation for industrial hygienists and IH labs available upon request
  • Analysis is not included
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Health & Safety Testing
Detectable Analytes Ethylene Oxide
Measurement Range 600 ppm/hr
Product Type Personal Monitor
Quantity 5/Cs.