Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethylene Oxide Solution

Catalog No. 779202100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
500 g
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-920-2100ML 1 L
77-920-21L 25 mL
77-920-225ML 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 77-920-2100ML Supplier Sigma Aldrich Supplier No. 779202100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethylene oxide solution can be used for the conversion of ortho-carborane to (2'sa-hydroxyethyl)-1,2-dicarba-closo-dodecaborane, a key step in the synthesis of ortho-carborane-modified N-alkyl-deoxynojirimycin (DNMs).

Specifications

CAS 75-21-8
Molecular Formula C2H4O
Linear Formula C2H4O
MDL Number MFCD00014482
Quantity 1 L
Molecular Weight (g/mol) 44.05 g/mol
Biological Activity Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.