Learn More
Ethylene Oxide Stock Solution, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™
Catalog No. 111001697 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1.2 mL
Catalog No. Quantity
11-100-1697 1.2 mL
1 options
Catalog No. 11-100-1697 Supplier MilliporeSigma Supelco Supplier No. PHR12951.2ML
Add to Cart
Add to Cart
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1.2 mL
Grade Certified Reference Material