Learn More
Medchemexpress LLC Ethylene-vinyl acetate copolymer | 24937-78-8 | MFCD00133996 | 40.0% | (C4H6O2.C2H4)x | 25 G
SDP
Catalog No. 5000280871
Encompass_Preferred
Ethylene-vinyl acetate copolymer (CAS 24937-78-8) is a research-grade copolymer used as a biochemical reagent or biological material in life-science research. The material is reported as a white to off-white liquid with a density of 0.948 g/cm³ and typical vinyl acetate content near 40 wt%. It is intended for laboratory and research applications where a vinyl-containing ethylene copolymer is required.
  • Research-grade copolymer for laboratory applications.
  • Suitable as a biological material or organic reagent in life-science studies.
  • White to off-white liquid form with reported density 0.948 g/cm³.
  • Typical vinyl acetate content approximately 40.0% (wt%).
  • Available in small lab-scale packages for controlled use.
Catalog No. 50-002-80871 Supplier Medchemexpress LLC Supplier No. HYW25012625G
Ethylene-vinyl acetate copolymer (CAS 24937-78-8) is a research-grade copolymer used as a biochemical reagent or biological material in life-science research. The material is reported as a white to off-white liquid with a density of 0.948 g/cm³ and typical vinyl acetate content near 40 wt%. It is intended for laboratory and research applications where a vinyl-containing ethylene copolymer is required.
  • Research-grade copolymer for laboratory applications.
  • Suitable as a biological material or organic reagent in life-science studies.
  • White to off-white liquid form with reported density 0.948 g/cm³.
  • Typical vinyl acetate content approximately 40.0% (wt%).
  • Available in small lab-scale packages for controlled use.