Learn More
Strem, An Ascensus Company CAS# 94-93-9. 25g. Ethylenebis(salicylimine), 98% SALEN. MFCD00002244
SDP
Supplier:  Strem, An Ascensus Company 07054025G
Encompass
CAS# 94-93-9. 25g. Ethylenebis(salicylimine), 98% SALEN. MFCD00002244. Molecular Weight: 268.32. Molecular Formula: HOC6H4CH=NCH2CH2N=CHC6H4OH. Color/form: yellow xtl. Strem# 07-0540. http://www.strem.com/catalog/v/07-0540/
Catalog No. 50-901-09016