Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediamine, (1,2-Diaminoethane), Reagents

Catalog No. 01337525
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-525 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-525 Supplier Reagents Holdings Llc Supplier No. C2120950500C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethylenediamine, (1,2-Diaminoethane), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

CAS 107-15-3
Boiling Range 116°C
Melting Point 8°C
Color Colorless to Slight Yellow
Boiling Point 116°C
Melting Range 8°C
Packaging Glass Bottle
Molecular Formula C2H8N2
Quantity 500 mL
Specific Gravity 0.898
Synonym 1,2-Ethanediamine, 1,2-Diaminoethane
InChI Key PIICEJLVQHRZGT-UHFFFAOYSA-N
SMILES NCCN
IUPAC Name ethane-1,2-diamine
Molecular Weight (g/mol) 60.10
Grade Reagent
Physical Form Liquid
Chemical Name or Material Ethylenediamine, (1,2-Diaminoethane)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.