Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediamine, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020554 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0554 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0554 Supplier MilliporeSigma Supelco Supplier No. 410081MLF
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 716°F
CAS 107-15-3
Vapor Pressure 10 mmHg (20°C)
Vapor Density 2.07 (vs. air)
Density 0.899 g/mL (at 25°C)
Flash Point 38°C
Physical Form Neat
Linear Formula NH2 CH2 CH2 NH2
MDL Number MFCD00008204
Melting Point 8.5°C
Boiling Point 118°C
Synonym 1,2-Diaminoethane
Type Analytical Standard
Quantity 1 mL
Formula Weight 60.10
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.