Learn More
AdipoGen Ethylenediamine disuccinate
SDP
Supplier:  AdipoGen CDXE0096G100
Encompass_Preferred
Chemical. CAS 20846-91-7. Formula C10H16N2O8. MW 292.24. Aminopolycarboxylic acid APCA. A powerful, readily biodegradable chelant. Complexing agent, exhibiting superior biological degradation environmentally friendly compared to EDTA. Used in cosmetic formulations, as effective anti-oxidant and water hardness control, to reduce radical generation, as effective peroxide stabiliser, as preservative potentiator improving piocide efficacy and excellent transition metal chelator. It forms stable water-soluble complexes with metal ions. It is a promising chelant for chelant-assisted phytoextraction of trace metals in polluted soil.
Catalog No. 50-256-0967