Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediamine Monohydrate 98.0+%, TCI America™
SDP

Catalog No. E0081500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0081500ML 500 mL
E008125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0081500ML Supplier TCI America Supplier No. E0081500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6780-13-8
Molecular Formula C2H10N2O
Molecular Weight (g/mol) 78.115
MDL Number MFCD00149563
InChI Key XZUAPPXGIFNDRA-UHFFFAOYSA-N
PubChem CID 12200817
IUPAC Name ethane-1,2-diamine;hydrate
SMILES C(CN)N.O

Specifications

Melting Point 10°C
Color Yellow
Boiling Point 118°C
Quantity 500 mL
UN Number 1604
Formula Weight 60.10
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Ethylenediamine Monohydrate
EINECSNumber (2)-0150
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.