Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethylenediamine-N,N'-Disuccinic Acid Trisodium Salt Solution

Catalog No. 926981L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
92-698-1L 100 mL
92-698-100ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 92-698-1L Supplier Sigma Aldrich Supplier No. 926981L
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethylenediamine-N,N'sa-disuccinic acid trisodium salt (EDDS) is an aminopolycarboxylic acid (APCA), which is used as a complexing agent. It forms stable water-soluble complexes with metal ions. EDDS has a better chelating property than EDTA. It can also be used as a flushing agent due to its eco-friendly characteristics and high biodegradability.powerful, readily biodegradable chelant

Specifications

CAS 121-45-9
Boiling Point 111°C to 112°C
Molecular Formula C10H13N2Na3O8
Linear Formula C10H13N2Na3O8
Quantity 100 mL
Synonym N,N'-Ethylenedi-(aspartic acid) trisodium salt; EDDS-Na3; Trisodium ethylenediamine-N,N'-disuccinate
Molecular Weight (g/mol) 358.19 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.