Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethylenediamine, polymer-bound 100-200 mesh, extent of labeling: 4.0-5.7 mmol/g loading, 1 % cross-linked | 177987-93-8 | MFCD00269799 | 5G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 4720935G
Encompass
Ethylenediamine, polymer-bound 100-200 mesh, extent of labeling: 4.0-5.7 mmol/g loading, 1 % cross-linked | 177987-93-8 | MFCD00269799 | 5G
Catalog No. 50-176-1236