Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethylenediamine puriss. p.
Catalog No. 502891471
Encompass
Ethylenediamine is a linear aliphatic diamine, widely used as a building block in organic synthesis. It readily forms heterocyclic imidazolidine derivatives, because of its bifunctional nature, having two amines. Ethylenediamine is also utilized as a raw material for the synthesis of chelating agents, polymers, agrochemicals and pharmaceutical intermediates.
Catalog No. 50-289-1471 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 035502.5L
Ethylenediamine is a linear aliphatic diamine, widely used as a building block in organic synthesis. It readily forms heterocyclic imidazolidine derivatives, because of its bifunctional nature, having two amines. Ethylenediamine is also utilized as a raw material for the synthesis of chelating agents, polymers, agrochemicals and pharmaceutical intermediates.