Learn More
Medchemexpress LLC Ethylenediaminetetra 1kg
SDP
Catalog No. 5000206936
Encompass_Preferred
Ethylenediaminetetraacetic acid (EDTA) sodium hydrate is a kind of metal chelating agent (binds to bivalent and trivalent metal cations including calcium) Ethylenediaminetetraacetic acid sodium hydrate has antibacterial anti-inflammatory antioxidant anti-hypercalcemia and anticoagulant activities Ethylenediaminetetraacetic acid sodium hydrate decreases the metal ion-catalyzed oxidative damage to proteins and allows maintenance of reducing environment during protein purification Ethylenediaminetetraacetic acid sodium hydrate can alleviate the liver fibrosis Ethylenediaminetetraacetic acid sodium hydrate can be used for coronary artery disease and neural system disease research[1][2][3][4][5][6][7]
Catalog No. 50-002-06936 Supplier Medchemexpress LLC Supplier No. HYW1057001KG
Ethylenediaminetetraacetic acid (EDTA) sodium hydrate is a kind of metal chelating agent (binds to bivalent and trivalent metal cations including calcium) Ethylenediaminetetraacetic acid sodium hydrate has antibacterial anti-inflammatory antioxidant anti-hypercalcemia and anticoagulant activities Ethylenediaminetetraacetic acid sodium hydrate decreases the metal ion-catalyzed oxidative damage to proteins and allows maintenance of reducing environment during protein purification Ethylenediaminetetraacetic acid sodium hydrate can alleviate the liver fibrosis Ethylenediaminetetraacetic acid sodium hydrate can be used for coronary artery disease and neural system disease research[1][2][3][4][5][6][7]