Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethylenediaminetetraacetic acid, 60-00-4, MFCD00003541, 100 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EDS100G
Encompass
Linear Formula: (HO2CCH2)2NCH2CH2N(CH2CO2H)2, Molecular Weight: 292.24, BioUltra, anhydrous, ≥99% (titration), mp: 250 °C, Synonym: (Ethylenedinitrilo)tetraacetic acid, EDTA, Edathamil, Ethylenedinitrilotetraacetic acid.
Catalog No. NC0345220